Learn More
Macherey-Nagel™ Chrombond™ FM 15/2 SiOH Polypropylene Flash Column
Product Code. 12884431 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
90 mm
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12884431 90 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12884431 Supplier Macherey-Nagel™ Supplier No. 730881
  • ONLINE EXCLUSIVE PRICE
    2605.00 NOK
    2463.83 NOK / Pack of 50
    Save 141.17 NOK
    5% Off
View all promotions
Estimated Shipment: 17-11-2026
For automated systems or vacuum manifolds
  • Automated systems or CHROMABOND™ vacuum manifolds
  • Polypropylene column, polyethylene filter element
  • High loading capacity and separation efficiency
  • Constant back pressure, airtight sealing
  • Standard Luer port For automated solid phase extraction, parallel flash purification/synthesis, high throughput screening/purification.
Specifications
Length (Metric) 90 mm
Material Polypropylene
Type Flash Column
Column Type Flash Column
For Use With (Equipment) Biotage FlashMaster Systems