Learn More
Sartorius Stedim Biotech™ Chromatography Papers, Sheets, Grade FN 5
Product Code. 15912800 Shop All Sartorius Products
Change view
Click to view available options
Unit Size:
Pack of 100
Product Code. unitSize
15912800 Pack of 100
1 options
Product Code. 15912800
Brand: Sartorius Stedim Biotech™ FT2505200200N
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Highly pure paper for blotting, chromatography, and a wide range of life science and diagnostics absorption applications.
  • Highly pure paper grade.
  • Ideal for blotting and chromatography.
  • Ideal for a wide range of absorption applications like those common in the life sciences and diagnostics.
  • Non-sterile, bulk packed.
TRUSTED_SUSTAINABILITY
Specifications
Description 100/Pk, 90 g/m2
Dimensions (L x W) 200 mm × 200 mm
Film Thickness 0.18 mm
Format Sheet
Grade FN 5
Material Cotton Linters (> 98 % alpha-cellulose; < 0.04 % ash)