Learn More
Sartorius Stedim Biotech™ Chromatography Papers, Discs, Grade FN 4
Product Code. 15902960 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric) Outer:
125 mm
300 mm
Unit Size:
Pack of 100
Product Code. Diameter (Metric) Outer unitSize
15902960 125 mm Pack of 100
15912960 300 mm Pack of 100
2 options
Product Code. 15902960
Brand: Sartorius Stedim Biotech™ FT3504125
Only null left
Add to basket
Add to basket
Highly pure paper for blotting, chromatography, and a wide range of life science and diagnostics absorption applications.
  • Highly pure paper grade.
  • Ideal for blotting and chromatography.
  • Ideal for a wide range of absorption applications like those common in the life sciences and diagnostics.
  • Non-sterile, bulk packed.
TRUSTED_SUSTAINABILITY
Specifications
Description 100/Pk, 125 g/m2
Diameter (Metric) Outer 125 mm
Film Thickness 0.24 mm
Format Disc
Grade FN 4
Material Cotton Linters (> 98 % alpha-cellulose; < 0.04 % ash)