Learn More
Sartorius Stedim Biotech™ Chromatography Paper, Roll, Grade FN 3
Product Code. 15922720 Shop All Sartorius Products
Change view
Click to view available options
Dimensions (L x W):
270 mm x 100 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15922720 270 mm x 100 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15922720 Supplier Sartorius Stedim Biotech™ Supplier No. FT15030270100
  • ONLINE EXCLUSIVE PRICE
    2725.00 NOK
    1745.45 NOK / Pack of 1
    Save 979.55 NOK
    35% Off
Estimated Shipment: 04-01-2027
Sartorius Stedim Biotech™ Chromatography Paper, Roll, Grade FN 3 is ideal for blotting, chromatography, and other absorption applications.
  • Highly pure paper grade.
  • Ideal for blotting and chromatography.
  • Ideal for a wide range of absorption applications like those common in the life sciences and diagnostics.
TRUSTED_SUSTAINABILITY
Specifications
Description Non-sterile, bulk packed reel, white
Dimensions (L x W) 270 mm x 100 m
Film Thickness 0.19 mm
For Use With (Application) Chromatography, blotting
Grade FN 3
Material Cotton Linters