Learn More
ROBU VitraPOR™ Chromatography-Column
Chromatography-Column with fused filter disc.
Brand:  ROBU VitraPOR™ 31102
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 16863086
  • 2355.00 NOK / Pack of 5
Estimated Shipment: 27-05-2024
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
5/Pk.
20 mL
Fused Filter Disc
2
Borosilicate Glass 3.3