Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and NS Socket
Product Code. 10529581
Click to view available options
Column Capacity:
8 mL
15 mL
23 mL
35 mL
125 mL
430 mL
Unit Size:
Each
Product Code. 10529581
Brand: Lenz Laborglasinstrumente™ 05450703
Only null left
Specifications
Description Chromatography Column
For Use With (Application) Chromatographic applications
Diameter (Metric) Inner 15 mm
Type Chromatography Column
Column Capacity 35 mL
Length (Metric) 200 mm
Material Duran Glass
Diameter (Metric) Outer 15 mm
Includes 0 to 2.5mm Bore PTFE-Hahn Stopcock