Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and NS Socket
Product Code. 10121811
Change view
Click to view available options
Column Capacity:
8 mL
15 mL
23 mL
35 mL
125 mL
430 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Column Capacity unitSize
10121811 125 mL Each
10030511 15 mL Each
10271171 23 mL Each
10668075 8 mL Each
10529581 35 mL Each
10191851 430 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10121811 Supplier Lenz Laborglasinstrumente™ Supplier No. 05450704
  • ONLINE EXCLUSIVE PRICE
    1325.00 NOK
    1018.14 NOK / Pack of 1
    Save 306.86 NOK
    23% Off
Estimated Shipment: 25-08-2026
Specifications
Description Chromatography Column
For Use With (Application) Chromatographic applications
Diameter (Metric) Inner 20 mm
Type Chromatography Column
Column Capacity 125 mL
Length (Metric) 400 mm
Material Duran Glass
Diameter (Metric) Outer 20 mm
Includes 0 to 2.5mm Bore PTFE-Hahn Stopcock