Learn More
Lenz Laborglasintrumente Chromatographic Columns with Frit and Beaded Rim
Product Code. 10413130
Click to view available options
Capacity (Metric) Reservoir:
8 mL
35 mL
430 mL
Unit Size:
Each
Product Code. 10413130
Brand: Lenz Laborglasintrumente 05445702
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Chromatographic columns with sintered frit (porosity: 0)
  • With PTFE stopcock
  • Made of DURAN™ tubing
Specifications
Capacity (Metric) Reservoir 35 mL
Length (Metric) 200 mm
Material Duran Glass
Diameter (Metric) Outer 15 mm