Learn More
Lenz Laborglasinstrumente™ Chromatographic Columns with Frit and Beaded Rim
Product Code. 10778461
Change view
Click to view available options
Column Capacity:
15 mL
125 mL
1000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Column Capacity Unit Size
10778461 125 mL Each
10693812 1000 mL Each
11984011 15 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10778461 Supplier Lenz Laborglasinstrumente™ Supplier No. 05445704
  • ONLINE EXCLUSIVE PRICE
    929.00 NOK
    811.94 NOK / Pack of 1
    Save 117.06 NOK
    12% Off
View all promotions
Estimated Shipment: 17-11-2026
Specifications
Description Chromatography Column
For Use With (Application) Chromatographic applications
Diameter (Metric) Inner 20 mm
Type Chromatography Column
Column Capacity 125 mL
Length (Metric) 400 mm
Material Duran Glass
Diameter (Metric) Outer 20 mm
Includes PTFE Stopcock