Learn More
Molecular Probes™ Chromatin Condensation & Membrane Permeability Dead Cell Apoptosis Kit with Hoechst 33342, YO-PRO™-1, and PI dyes, for flow cytometry
Product Code. 10424202
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10424202 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10424202 Supplier Molecular Probes™ Supplier No. V23201
  • ONLINE EXCLUSIVE PRICE
    5790.00 NOK
    4458.30 NOK / Pack of 1
    Save 1331.70 NOK
    23% Off
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Vybrant™ Apoptosis Assay Kit #7 - Hoechst 33342/YO-PRO™-1/propidium iodide - 200 assays
This product detects apoptotic cells with changes in nuclear chromatin condensation and plasma membrane permeability, using three nucleic acid stains: UV-excitable Hoechst 33342, green fluorescent YO-PRO™ dye, and propidium iodide. The YO-PRO™ dye can enter apoptotic cells, whereas red-fluorescent propidium iodide (PI) cannot. Thus after staining with YO-PRO™-1 dye and PI, apoptotic cells show green fluorescence, and dead cells show primarily red fluorescence and some green fluorescence. Blue fluorescent Hoechst 33342 brightly stains the condensed chromatin of apoptotic cells and more dimly stains the normal chromatin of live cells. The staining pattern resulting from the simultaneous use of these three dyes makes it possible to distinguish normal, apoptotic and dead cell populations by flow cytometry or fluorescence microscopy.
View a selection guide for all apoptosis assays for flow cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Description Chromatin Condensation and Membrane Permeability Dead Cell Apoptosis Kit, 1 Kit, 200 rxns, Tube Format, 461nm Emission (Hoechst 33342), 350nm Excitation (Hoechst 33342), 617nm Emission (PI), 535nm Excitation (PI), 509nm Emission (YO-PRO™-1), 491nm Excitation (YO-PRO™-1), 2 to 8°C Storage Temperature, Water Soluble
Quantity 1 Kit
Format Tube
Storage Buffer 0.2M Hydrochloric acid/Methanol (1:500 dilution), Methanol (1:10 dilution), 50mM phosphate buffer, 7pH (1:20 dilution)
Product Type Chromatin Condensation and Membrane Permeability Dead Cell Apoptosis Kit
Content And Storage 1 vial of Hoechst 33342, 1 vial of YO-PRO™-1, and propidium iodide. Store at 2°C to 8°C
For Use With (Application) Cell Viability, Cellular Imaging, Flow Cytometry, Pharms and Biopharma, Proliferation and Function, Target-based ADME/Tox Assays
Conjugate Hoechst 33342, Propidium Iodide, YO-PRO-1