missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab CHROMagar™ Orientation
Description
- Chromogenic dehydrated medium for isolation and differentation of urinary tract pathogens.
- It can also be used as a general nutrient agar for the isolation of pathogens.
Specifications
Specifications
| Form | Powder |
| Media Type | CHROMagar Orientation |
| Product Type | Microbiology Media |
| Formulation | Agar 15.0 g/L, Peptone and Yeast Extract 17.0 g/L, Chromogenic mix 1.0 g/L |
| Incubation Conditions | 37°C |
| Incubation Time | 18 to 24 hr. |
| pH | 7.0 ±0.2 |
| Yield | 5 L |
| Weight (Metric) | 165 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?