Learn More
Macherey-Nagel™ Chromafix™ SB Cartridge
Product Code. 10236571 Shop All Macherey-Nagel Products
Change view
Click to view available options
Description:
Adsorbent Weight: 230 mg
Adsorbent Weight: 460 mg
Adsorbent Weight: 920 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10236571 Adsorbent Weight: 920 mg Pack of 50
10458981 Adsorbent Weight: 230 mg Pack of 50
10053051 Adsorbent Weight: 460 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10236571 Supplier Macherey-Nagel™ Supplier No. 731836
  • ONLINE EXCLUSIVE PRICE
    3390.00 NOK
    3113.39 NOK / Pack of 50
    Save 276.61 NOK
    8% Off
In Stock
Specifications
Base Material Silica
Description Adsorbent Weight: 920 mg
For Use With (Application) Organic Acids, Caffeine, Saccharin
Particle Size 45 μm
Sorbent SB
Type Chromafix SB Cartridge
Pore Size 60 Å
Quantity 50 Pack
Surface Area (Metric) 500 m2/g