Learn More
Macherey-Nagel™ Chromafix™ SA Cartridge
Product Code. 10175041 Shop All Macherey-Nagel Products
Change view
Click to view available options
Description:
Adsorbent Weight: 220 mg
Adsorbent Weight: 450 mg
Adsorbent Weight: 920 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10175041 Adsorbent Weight: 220 mg Pack of 50
10135091 Adsorbent Weight: 450 mg Pack of 50
10145091 Adsorbent Weight: 920 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10175041 Supplier Macherey-Nagel™ Supplier No. 731831
  • ONLINE EXCLUSIVE PRICE
    2120.00 NOK
    2004.66 NOK / Pack of 50
    Save 115.34 NOK
    5% Off
View all promotions
Estimated Shipment: 02-12-2026
Specifications
Base Material Silica
Description Adsorbent Weight: 220 mg
For Use With (Application) Amino Acids, Amines, Chlorophyll, PCBs
Particle Size 45 μm
Sorbent SA
Type Chromafix SA Cartridge
Pore Size 60 Å
Quantity 50 Pack
Surface Area (Metric) 500 m2/g