Learn More
Macherey-Nagel™ Chromafix™ PS-RP cartridges
Product Code. 10105141 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10105141 50 Pack Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10105141 Supplier Macherey-Nagel™ Supplier No. 731875
  • ONLINE EXCLUSIVE PRICE
    2950.00 NOK
    2788.61 NOK / Pack of 50
    Save 161.39 NOK
    5% Off
In Stock
Specifications
Base Material Hydrophobic PS/DVB Copolymer
For Use With (Application) Removal of Interfering Organic Components From Water
Particle Size 100 μm
Type Chromafix PS-RP Cartridge
Pore Size 100 Å
Quantity 50 Pack