Learn More
Macherey-Nagel™ Chromafix™ HR-XCW Cartridge
Product Code. 12767740 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
155 mg
240 mg
500 mg
Unit Size:
Pack of 50
Product Code. Bed Weight unitSize
12767740 500 mg Pack of 50
12757740 240 mg Pack of 50
12798252 155 mg Pack of 50
3 options
Product Code. 12767740
Brand: Macherey-Nagel™ 731776
Only null left
Specifications
Base Material Spherical PS/DVB Copolymer
Bed Weight 500 mg
For Use With (Application) Basic Compounds Like Quaternary Amines, Basic Active Ingredients From Strongly Matrix-Contaminated Samples
Particle Size 45 or 85 μm
Sorbent Weak carboxylic acid modified polymeric cation exchanger
Type Chromafix HR-XCW Cartridge
Pore Size 50 to 60 Å
Quantity 50 Pack
Surface Area (Metric) 850 m2/g