Learn More
Macherey-Nagel™ Chromafix™ HR-XCW Cartridge
Product Code. 12757740 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
155 mg
240 mg
500 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight unitSize
12757740 240 mg Pack of 50
12767740 500 mg Pack of 50
12798252 155 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12757740 Supplier Macherey-Nagel™ Supplier No. 731775
Estimated Shipment: 28-08-2026
Specifications
Base Material Spherical PS/DVB Copolymer
Bed Weight 240 mg
For Use With (Application) Basic Compounds Like Quaternary Amines, Basic Active Ingredients From Strongly Matrix-Contaminated Samples
Particle Size 45 or 85 μm
Sorbent Weak carboxylic acid modified polymeric cation exchanger
Type Chromafix HR-XCW Cartridge
Pore Size 50 to 60 Å
Quantity 50 Pack
Surface Area (Metric) 850 m2/g