Learn More
Macherey-Nagel™ Chromafix™ HR-XC Cartridge
Product Code. 12894301 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
155 mg
240 mg
500 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
12894301 500 mg Pack of 50
12830961 155 mg Pack of 50
12801041 240 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12894301 Supplier Macherey-Nagel™ Supplier No. 731757
  • ONLINE EXCLUSIVE PRICE
    3905.00 NOK
    3695.75 NOK / Pack of 50
    Save 209.25 NOK
    5% Off
Estimated Shipment: 02-12-2026
Specifications
Base Material Spherical PS/DVB Copolymer
Bed Weight 500 mg
For Use With (Application) Basic Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum, Fungicides From Food
Particle Size 45 or 85 μm
Sorbent Benzenesulfonic acid modified polymeric cation exchanger
Type Chromafix HR-XC Cartridge
Pore Size 65 to 75 Å
Quantity 50 Pack
Surface Area (Metric) 800 m2/g