Learn More
Macherey-Nagel™ Chromafix™ HR-XC Cartridge
Product Code. 12830961 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
155 mg
240 mg
500 mg
Unit Size:
Pack of 50
Product Code. Bed Weight unitSize
12830961 155 mg Pack of 50
12894301 500 mg Pack of 50
12801041 240 mg Pack of 50
3 options
Product Code. 12830961
Brand: Macherey-Nagel™ 731755
Only null left
Specifications
Base Material Spherical PS/DVB Copolymer
Bed Weight 155 mg
For Use With (Application) Basic Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum, Fungicides From Food
Particle Size 45 or 85 μm
Sorbent Benzenesulfonic acid modified polymeric cation exchanger
Type Chromafix HR-XC Cartridge
Pore Size 65 to 75 Å
Quantity 50 Pack
Surface Area (Metric) 800 m2/g