Learn More
Macherey-Nagel™ Chromafix™ HR-XAW Cartridge
Product Code. 12788252 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
155 mg
240 mg
500 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
12788252 500 mg Pack of 50
12778252 240 mg Pack of 50
12768252 155 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12788252 Supplier Macherey-Nagel™ Supplier No. 731773
  • ONLINE EXCLUSIVE PRICE
    3905.00 NOK
    3695.75 NOK / Pack of 50
    Save 209.25 NOK
    5% Off
Estimated Shipment: 02-12-2026
Specifications
Base Material Spherical PS/DVB Copolymer
Bed Weight 500 mg
For Use With (Application) Perfluorinated Surfactants, Acidic Compounds Like Sulfonates, Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum
Particle Size 45 or 85 μm
Sorbent Weakly basic secondary and tertiary ammonium polymeric anion exchanger
Type Chromafix HR-XAW Cartridge
Pore Size 55 to 65 Å
Quantity 50 Pack
Surface Area (Metric) 850 m2/g