Learn More
Macherey-Nagel™ Chromafix™ HR-XA Cartridge
Product Code. 12824371 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
155 mg
240 mg
500 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
12824371 155 mg Pack of 50
12804441 240 mg Pack of 50
12814511 500 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12824371 Supplier Macherey-Nagel™ Supplier No. 731768
  • ONLINE EXCLUSIVE PRICE
    2750.00 NOK
    2598.24 NOK / Pack of 50
    Save 151.76 NOK
    5% Off
Estimated Shipment: 02-12-2026
Specifications
Base Material Spherical PS/DVB Copolymer
Bed Weight 155 mg
For Use With (Application) Acidic Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum, Phenolic Acids, Acidic Herbicides
Particle Size 45 or 85 μm
Sorbent Strong quat. ammonium modified polymeric anion exchanger
Type Chromafix HR-XA Cartridge
Pore Size 55 to 65 Å
Quantity 50 Pack
Surface Area (Metric) 850 m2/g