Learn More
Macherey-Nagel™ Chromafix™ HR-P Cartridge
Product Code. 10678572 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
200 mg
330 mg
680 mg
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Weight Unit Size
10678572 200 mg Pack of 50
10368211 330 mg Pack of 50
10246851 680 mg Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10678572 Supplier Macherey-Nagel™ Supplier No. 731839
  • ONLINE EXCLUSIVE PRICE
    2180.00 NOK
    2060.66 NOK / Pack of 50
    Save 119.34 NOK
    5% Off
View all promotions
Estimated Shipment: 02-12-2026
Resolve a number of components from water or oil. Chromafix™ HR-P Cartridges contain highly porous polystyrene-divinylbenzene adsorbent resin for SPE.
Features
  • Highly porous polystyrene-divinylbenzene copolymer
  • Specific surface 1200 m2/g
  • Particle size 50–100 μm
  • Very high binding capacity, up to 30 percent of adsorbent weight (for comparison: silica adsorbents about 3 percent)
Recommended application:
  • Aromatic compounds
  • Phenols from water
  • Nitroaromatics from water
  • Pesticides from water
  • PAHs from oil
Specifications
Bed Weight 200 mg
For Use With (Application) Aromatic Compounds, Phenols From Water, Nitroaromatics From Water, Pesticides From Water, PAHs From Oil
Particle Size 50 to 100 μm
Sorbent HR-P
Type Chromafix HR-P Cartridge
Quantity 50 Pack
Resin Composition Polystyrene-divinylbenzene adsorbent resin
Surface Area (Metric) 1200 m2/g