Learn More
Macherey-Nagel™ Chromafix™ C4 Cartridge
Product Code. 10092821 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10092821 50 Pack Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10092821 Supplier Macherey-Nagel™ Supplier No. 731741
  • ONLINE EXCLUSIVE PRICE
    2380.00 NOK
    2251.06 NOK / Pack of 50
    Save 128.94 NOK
    5% Off
View all promotions
In Stock
Specifications
Base Material Silica
Carbon Load 7%
For Use With (Application) Compounds, Which Are Too Strongly Retained on C18 or C8
Particle Size 45 μm
Type Chromafix C4 Cartridge
Endcapped No
Pore Size 60 Å
Quantity 50 Pack
Surface Area (Metric) 500 m2/g