Learn More
Macherey-Nagel™ Chromafix™ Alox N SPE Cartridge
Product Code. 10516351 Shop All Macherey-Nagel Products
Change view
Click to view available options
Bed Weight:
850 mg
1700 mg
Unit Size:
Pack of 50
Product Code. Bed Weight unitSize
10516351 1700 mg Pack of 50
10134801 850 mg Pack of 50
2 options
Product Code. 10516351
Brand: Macherey-Nagel™ 731845
Specifications
Bed Weight 1700 mg
For Use With (Application) Solid Phase Extraction
Sorbent Chromafix Alox N
Type SPE Cartridge