Learn More
Thermo Scientific™ Chromacol Vial Racks
Product Code. 10759114
Change view
Click to view available options
Holds:
Vials
Vials (100)
Vials (104)
Vials (105)
Material:
Aluminum
Foam
Plastic
For Use With (Equipment):
11 mm OD Vials
12 mm OD Vials
15 mm OD Vials
22 to 23mm OD Vials
22-23 mm OD Vials
8 mm OD Vials
Unit Size:
Each
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Material For Use With (Equipment) Unit Size
10759114 Vials (100) Plastic 12 mm OD Vials Pack of 5
11922585 Vials Aluminum 12 mm OD Vials Each
10235812 Vials Aluminum 22-23 mm OD Vials Pack of 5
10162882 Vials Aluminum 15 mm OD Vials Pack of 5
10494302 Vials (104) Aluminum 11 mm OD Vials Pack of 5
10122932 Vials (105) Foam 12 mm OD Vials Pack of 5
10529922 Vials Foam 8 mm OD Vials Pack of 5
10769304 Vials Plastic 22 to 23mm OD Vials Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10759114 Supplier Thermo Scientific™ Supplier No. B100
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 26-11-2026
Ensure a safe working position on the lab bench and during transport with these robust vial racks.
Save space in the fridge while ensuring a safe working position on the bench and during transport. Available in a various materials, colors and capacities, Thermo Scientific™ Chromacol Vial Racks meet a variety of application needs
Choose from an assortment of chemically resistant and fairly robust racks.
  • Aluminum and plastic models are temperature resistant from -90 to +121°C (-130 to +250°F)
  • Aluminum and plastic models are autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Description Vial Racks
Autoclavable Autoclavable
Color Assorted
Holds Vials (100)
Material Plastic
For Use With (Equipment) 12 mm OD Vials
Quantity 5
Product Type Vial Rack
Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.