Learn More
Macherey-Nagel™ Chromabond™ Tubing Adapter
Product Code. 12860771 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
4 Pk.
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12860771 4 Pk. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12860771 Supplier Macherey-Nagel™ Supplier No. 730387
  • ONLINE EXCLUSIVE PRICE
    3130.00 NOK
    2878.21 NOK / Pack of 4
    Save 251.79 NOK
    8% Off
Estimated Shipment: 16-12-2026
For use with vacuum manifolds
CHROMABOND™ vacuum manifold for simultaneous preparation of up to 12 samples. The manifold consists of a rectangular glass cabinet with a vacuum gauge and a polypropylene lid, that can hold the columns or cartridges. The replaceable valves on the lid allow individual vacuum control for each solid phase extraction column, if required. The cabinet is fitted with a variable rack with exchangeable partitions, that accept a wide variety of vessels like test tubes, measuring flasks, scintillation vials, autosampler vials, plastic vials and many more. The vacuum in the chamber can be adjusted with the control valve and read from the gauge.
Specifications
For Use With (Equipment) 3 and 6 mL Glass Columns, Application of Large Sample Volumes
Type Tubing Adapter
Quantity 4 Pk.