Specifications
| Base Material | Silica |
| Column Capacity (Metric) | 3 mL |
| Column Type | SPE |
| Bed Weight | 200 mg |
| Particle Size | 45 μm |
| Sorbent | Chromabond SiOH |
| Type | Solid Phase Extraction Column |
| Pore Size | 60 Å |
| Surface Area (Metric) | 500 m2/g |