Specifications
| Column Capacity (Metric) | 3 mL |
| Column Type | SPE |
| Bed Weight | 500 mg |
| Particle Size | 45 μm |
| Type | Solid Phase Extraction Column |
| Pore Size | 60 μm |
| Surface Area (Metric) | 500 m2/g |