Learn More
Macherey-Nagel™ Chromabond™ SA Polypropylene SPE Column
Brand: Macherey-Nagel™ 730077
Additional Details : Weight : 0.01000kg
Specifications
Silica | |
SPE | |
45 μm | |
60 μm |
3 mL | |
500 mg | |
Solid Phase Extraction Column | |
500 m2/g |