Learn More
Macherey-Nagel™ Chromabond™ PTL Separation Column
Product Code. 12864431 Shop All Macherey-Nagel Products
Click to view available options
Column Capacity (Metric):
1 mL
3 mL
30 mL
45 mL
70 mL
Unit Size:
Pack of 100
Pack of 50
Product Code. 12864431
Brand: Macherey-Nagel™ 730740
Only null left
Add to basket
Add to basket
Separate compounds with gravity flow phase separation. Chromabond™ PTL Separation Columns come in a variety of capacities
  •  Automatic separation of a two-phase mixture without separation funnel
  • Two-phase mixtures are completely applied to the column and the phase boundary is determined without further work.
  • The special membrane automatically stops the flow when the lower phase has passed.
  • The upper phase remains in the column, thus both phases are available for futher analysis. 
  • Columns must not be run with vacuum or pressure
  • PTL for solvents lighter than water, e.g., for diethyl ether, hexane
  • maximum size 70 mL
Specifications
Column Capacity (Metric) 45 mL
Column Type SPE
For Use With (Application) For Solvents Lighter Than Water, Diethyl Ether, Hexane
Sorbent Chromabond PTL
Type Phase Separation Column