Learn More
Macherey-Nagel™ Chromabond™ Polyethylene Flash DL Cartridge
Product Code. 15333491 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
171 mm
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15333491 171 mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15333491 Supplier Macherey-Nagel™ Supplier No. 732983
  • ONLINE EXCLUSIVE PRICE
    3175.00 NOK
    3001.41 NOK / Pack of 20
    Save 173.59 NOK
    5% Off
View all promotions
In Stock
Specifications
Length (Metric) 171 mm
Material Polyethylene
Type Flash DL Cartridge
Diameter (Metric) Outer 26.4 mm