Learn More
Chroma Gesellschaft™ Heat Resistant Bench Mats
Product Code. 12777009
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
150 x 150 mm
225 x 225 mm
300 x 300 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. Dimensions (L x W) unitSize
12777009 225 x 225 mm Pack of 10
10466412 150 x 150 mm Pack of 10
10520814 300 x 300 mm Pack of 5
3 options
Product Code. 12777009
Brand: FGF Aston BOR600030K
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Hard, heat resistant squares, 6mm thick, consisting of reinforced calcium silicate matrix. Not suitable for direct heating applications. Should mats become wet, allow them to dry gradually at ambient temperature.
Specifications
Product Type Microplate
Material Polypropylene
Dimensions (L x W) 225 x 225 mm
Thickness 6 mm
Quantity 100Pack