missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18037-1-AP-150UL
This item is not returnable.
View return policy
Description
The nicotinic acetylcholine receptors are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are stated that the proposed structure for each subunit is a conserved N-terminal extracellular domain followed by 3 conserved transmembrane domains, a variable cytoplasmic loop, a fourth conserved transmembrane domain, and a short C-terminal extracellular region.Specifications
| CHRNB3 | |
| Polyclonal | |
| Unconjugated | |
| CHRNB3 | |
| CHRNB3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108043, 1142, 171131 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12391, Q05901, Q8BMN3 | |
| Chrnb3 | |
| CHRNB3 Fusion Protein Ag12664 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction