missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11388-1-AP-150UL
This item is not returnable.
View return policy
Description
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.Specifications
| CHRNA6 | |
| Polyclonal | |
| Unconjugated | |
| CHRNA6 | |
| CHNRA6, CHRNA6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11440, 81721, 8973 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P43143, Q15825, Q9R0W9 | |
| CHRNA6 | |
| CHRNA6 Fusion Protein Ag1921 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction