missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10477-1-AP-20UL
This item is not returnable.
View return policy
Description
CHMP2A belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A is required for both MVB formation and regulation of cell cycle progression.Specifications
| CHMP2A | |
| Polyclonal | |
| Unconjugated | |
| CHMP2A | |
| CHMP2, CHMP2A, chromatin modifying protein 2A, hVps2 1, VPS2, Vps2 1, VPS2A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27243, 365191, 68953 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43633, Q9DB34 | |
| CHMP2A | |
| CHMP2A Fusion Protein Ag0719 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction