Learn More
Chlorphenoxamine, MedChemExpress
Product Code. 16454617
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454617 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16454617 Supplier Medchem Express Supplier No. HYB1607
MedChemExpress Chlorphenoxamine is an antihistamine and anticholinergic used as an antipruritic and antiparkinsonian agent.
Chlorphenoxamine is an antihistamine and anticholinergic used as an antipruritic and antiparkinsonian agent.
Specifications
Chemical Name or Material Chlorphenoxamine
CAS 77-38-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H22ClNO
Quantity 50 mg
Solubility Information DMSO : ≥ 48 mg/mL (157.98 mM)
SMILES CN(CCOC(C1=CC=CC=C1)(C2=CC=C(C=C2)Cl)C)C
Molecular Weight (g/mol) 303.83
Formula Weight 303.83
Percent Purity 95.09%
Grade Research
Physical Form Oil
For Use With (Application) Neuroscience-Neurodegeneration