Learn More
Chlorpheniramine, TRC
Product Code. 30134893
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134893 50 mg 50mg
30133872 25 mg 25mg
30012639 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134893 Supplier Toronto Research Chemicals Supplier No. TRCC42470550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 132-22-9
Molecular Formula C16 H19 Cl N2
Molecular Weight (g/mol) 274.79
Synonym 2-Pyridinepropanamine, γ-(4-chlorophenyl)-N,N-dimethyl-, Pyridine, 2-[p-chloro-α-[2-(dimethylamino)ethyl]benzyl]- (8CI), γ-(4-Chlorophenyl)-N,N-dimethyl-2-pyridinepropanamine, (±)-Chloropheniramine, (±)-Chlorpheniramine, 1-(p-Chlorophenyl)-1-(2-pyridyl)-3-dimethylaminopropane, 2-[p-Chloro-α-[2-(dimethylamino)ethyl]benzyl]pyridine, 3-(p-Chlorophenyl)-3-(2-pyridyl)-N,N-dimethylpropylamine, 4-Chloropheniramine, Allergican, Chloropheniramine, Chlorophenylpyridamine, Chloroprophenpyridamine, Chlorphenamine, Chlorpheniramine, Chlorprophenpyridamine, Haynon, RS-Chlorpheniramine, dl-1-(p-Chlorophenyl)-1-(2-pyridyl)-3-(dimethylamino)propane, γ-(4-Chlorophenyl)-γ-(2-pyridyl)propyldimethylamine
IUPAC Name 3-(4-chlorophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine
SMILES CN(C)CCC(c1ccc(Cl)cc1)c2ccccn2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C16H19ClN2/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13/h3-9,11,15H,10,12H2,1-2H3
Formula Weight 274.1237
Percent Purity >95%
Chemical Name or Material Chlorpheniramine
Grade HPLC