missing translation for 'onlineSavingsMsg'
Learn More

Chlorothiazide, TRC

Product Code. 17924815
Change view
Click to view available options
Quantity:
50 mg
250 mg
2.5 g
Unit Size:
2.5g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924815 50 mg 50mg
30134301 2.5 g 2.5g
17924585 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17924815 Supplier Toronto Research Chemicals Supplier No. TRCC38000050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-94-6
Formula Weight 294.9488
InChI Formula InChI=1S/C7H6ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-3H,(H,10,11)(H2,9,12,13)
IUPAC Name 6-chloro-1,1-dioxo-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
Molecular Formula C7 H6 Cl N3 O4 S2
Molecular Weight (g/mol) 295.72
Percent Purity >95%
SMILES NS(=O)(=O)c1cc2c(cc1Cl)N=CNS2(=O)=O
Synonym 6-Chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 6-Chloro-7-sulfamoyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 6-Chloro-7-sulfamyl-1,2,4-benzothiadiazine 1,1-dioxide, Alurene, Chlorosal, Chlorothiazid, Chlorothiazide, Chlorthiazide, Chlorurit, Chlotride, Clotride, Diuresal, Diuril, Diuril Boluses, Diurilix, Diurite, Diutrid, Flumen, Minzil, NSC 25693, Neo-Dema, Salisan, Salunil, Saluretil, Saluric, Thiazide, Urinex, Warduzide, Yadalan
Chemical Name or Material Chlorothiazide
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.