missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Supelco™

Product Code. 15854250
Change view
Click to view available options
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15854250 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15854250 Supplier Supelco Supplier No. 1024322500
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ100 Graded - For Non-regulated Laboratory Applications
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For gas chromatography ECD and FID SupraSolv™

SupraSolv™ ECD and FID is a high-purity solvent grade specifically formulated for sample preparation in gas chromatography with electron capture (ECD) and flame ionization (FID) detection.ECD and FID impose distinct and stringent solvent purity requirements. The electron capture detector is exquisitely sensitive to electronegative species — particularly halogenated compounds — at sub-nanogram concentrations, while the flame ionization detector responds to any carbon-containing organic impurity undergoing combustion. SupraSolv™ ECD and FID solvents are therefore subjected to rigorous purification and detector-specific testing protocols to ensure that intrinsic solvent impurities remain below the respective detection thresholds of both detector types simultaneously.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.492 g/mL at 25°C
Molecular Weight (g/mol) 119.38
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.