missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Supelco™

Product Code. 15867110
Change view
Click to view available options
Unit Size:
1 L
10 L
2.5 L
25 L
Quantity:
1 L
10 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Quantity
15867110 25 L -
15804260 1 L 1 L
15814260 2.5 L -
15857110 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15867110 Supplier Supelco Supplier No. 1024456025
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For analysis EMSURE™ ACS,ISO,Reag. Ph Eur

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.492 g/mL at 25°C
Molecular Weight (g/mol) 119.38
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.