missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Supelco™

Product Code. 15816730
Change view
Click to view available options
Unit Size:
1 L
2.5 L
Quantity:
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Quantity
15816730 2.5 L -
15806730 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15816730 Supplier Supelco Supplier No. 8222652500
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

EMPLURA™

EMPLURA™ is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.492 g/mL at 25°C
Molecular Weight (g/mol) 119.38
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.