missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform (Anhydrous) Contains Amylenes as stabilizer, TRC
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Chemical Name or Material | Chloroform |
| CAS | 67-66-3 |
| Molecular Formula | C H Cl3 |
| Quantity | 100 mL |
| Synonym | Methane, trichloro-, Chloroform (8CI), Trichloromethane, F 20, HCC 20, NSC 77361, R 20, R 20 (refrigerant), Trichloroform |
| InChI Formula | InChI=1S/CHCl3/c2-1(3)4/h1H |
| SMILES | ClC(Cl)Cl |
| IUPAC Name | chloroform |
| Molecular Weight (g/mol) | 119.38 |
| Formula Weight | 117.9144 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction