Learn More
Chloroform 5000 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30312483
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312483 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30312483 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011074ME
 Restricted Item
Estimated Shipment: 30-12-2026
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mL
Concentration 5000 μg/mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol