missing translation for 'onlineSavingsMsg'
Learn More

βChloro-L-Alanine, MP Biomedicals

Product Code. 11470916 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11470916

Brand: MP Biomedicals 0520206780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 2731-73-9
Molecular Formula C3H6ClNO2
Quantity 100 mg
Synonym 3-chloro-l-alanine, r-2-amino-3-chloropropanoic acid, alanine, 3-chloro, beta-chloro-l-alanine, h-ala cl-oh, 2r-2-amino-3-chloropropanoic acid, l-alanine, 3-chloro, 2r-beta-chloroalanine, alanine, 3-chloro-, l, snxhmvhai jihl@
InChI Key ASBJGPTTYPEMLP-REOHCLBHSA-N
SMILES C(C(C(=O)O)N)Cl
IUPAC Name (2R)-2-amino-3-chloropropanoic acid
Molecular Weight (g/mol) 123.536
PubChem CID 439772
ChEBI CHEBI:17403
Formula Weight 123.6
Chemical Name or Material β-Chloro-L-Alanine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.