Learn More
chloro(diethyl)phosphine, 95%
Product Code. p-3735955 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. 10255771
Brand: Thermo Scientific Acros 350310010
Request Bulk or Custom Format
CAS: 686-69-1 | C4H10ClP | 124.55 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 686-69-1
Molecular Formula C4H10ClP
Molecular Weight (g/mol) 124.55
MDL Number MFCD00013621
InChI Key INJBDKCHQWVDGT-UHFFFAOYSA-N
Synonym chlorodiethylphosphine, diethylchlorophosphine, chloro diethyl phosphine, phosphinous chloride, diethyl, pubchem6480, chloro diethyl phosphane, acmc-1avkn, diethylphosphinous chloride, diethylphosphinous chloride #, chlorodiethyl phosphine
PubChem CID 69639
IUPAC Name chloro(diethyl)phosphane
SMILES CCP(CC)Cl
Specifications
Color Colorless to Yellow
Density 1.0230g/mL
Boiling Point 131.0°C to 132.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3CH2)2PCl
Refractive Index 1.4740 to 1.4760
Quantity 1 g
Specific Gravity 1.023
Formula Weight 124.55
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Chloro(diethyl)phosphine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloro(diethyl)phosphine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.