Learn More
Chlormezanone, TRC
Specifications
Specifications
| Chemical Name or Material | Chlormezanone |
| CAS | 80-77-3 |
| Molecular Formula | C11 H12 Cl N O3 S |
| Quantity | 100 mg |
| Synonym | Chlormezanone, 4H-1,3-Thiazin-4-one, 2-(4-chlorophenyl)tetrahydro-3-methyl-, 1,1-dioxide, 4H-1,3-Thiazin-4-one, 2-(p-chlorophenyl)tetrahydro-3-methyl-, 1,1-dioxide (6CI,7CI,8CI), (±)-Chlormezanone, (±)-Fenarol, 2-(4-Chlorophenyl)-3-methyl-4-metathiazanone-1,1-dioxide, 2-(p-Chlorophenyl)-3-methyl-1,3-perhydrothiazin-4-one 1,1-dioxide, 2-(p-Chlorophenyl)tetrahydro-3-methyl-4H-1,3-thiazin-4-one 1,1-dioxide, 2-(p-Chlorphenyl)-3-methyl-1,3-perhydrothiazin-4-on-1,1-dioxide, Alinam, Banabin-Sintyal, Chlormethazanone, Chlormethazone, Dichloromezanone, Fenarol, Lobak, Mio-Sed, Muskel Trancopal, NSC 169108, Phenarol, Rexan, Rilansyl, Rilaquil, Rilasol, Rilassol, Supotran, Suprotran, Tanafol, Trancopal, Trancote, Transanate, dl-Chlormezanone |
| InChI Formula | InChI=1S/C11H12ClNO3S/c1-13-10(14)6-7-17(15,16)11(13)8-2-4-9(12)5-3-8/h2-5,11H,6-7H2,1H3 |
| SMILES | CN1C(c2ccc(Cl)cc2)S(=O)(=O)CCC1=O |
| IUPAC Name | 2-(4-chlorophenyl)-3-methyl-1,1-dioxo-1,3-thiazinan-4-one |
| Molecular Weight (g/mol) | 273.74 |
| Formula Weight | 273.0226 |