Learn More
Chloride Reagent Set, Lovibond™
Click to view available options
Quantity:
75 Tests
Unit Size:
100 tests
Specifications
Specifications
| Quantity | 75 Tests |
| Type | Chloride Reagent Set |
Safety and Handling
Product Identifier
- reagent test chloride
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H290-May be corrosive to metals.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- MIXTURE LIST-Contains : iron(III) nitrate nonahydrate, nitric acid, ethane-1,2-diol,mercury dithiocyanate