Learn More
Palintest™ Chloride (Chloridol) Test Kit
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
Description
- Test kit for measuring Chloride (Chloridol)
- Range of 0 to 50,000 (NaCl) mg/L
- X50 Test Chloride 0-50 PPM
Specifications
Specifications
| No. of Tests | 50 |
| Quantity | 50 Pack |
| Range | 0 to 50,000mg/L |
| Sufficient For | 50 Tests |
| Type | Chloride (Chloridol) Test Kit |