Learn More
Chlorhexidine diacetate 100 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Chlorhexidine diacetate |
| CAS | 56-95-1 |
| Analyte or Component Names | Chlorhexidine diacetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H30 Cl2 N10 . 2 C2 H4 O2 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Chlorhexidine Diacetate, N,N''-Bis(4-chlorophenyl)-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide acetate (1:2), N,N''-Bis(4-chlorophenyl)-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide diacetate, 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide diacetate, 1,6-Bis(p-chlorophenylbiguanido)hexane diacetate, Arlacide A, Bactigras, Chlorasept 2000, Chlorhexidine acetate, Chlorhexidine diacetate, Chlorzoin, Dosisepsine, EC 40, EC 40 (antibacterial), Hibitane diacetate, Jie-Yin Liquid Disinfectant, NSC 526936 |
| InChI Formula | InChI=1S/C22H30Cl2N10.2C2H4O2/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18;2*1-2(3)4/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34);2*1H3,(H,3,4) |
| SMILES | CC(=O)O.CC(=O)O.Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1 |
| Show More |