missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloranilic acid, 100.7%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| CAS | 87-88-7 |
| Color | Red to Orange |
| Molecular Formula | C6H2Cl2O4 |
| Quantity | 25 g |
| Synonym | chloranilic acid, 2,5-dichloro-3,6-dihydroxy-p-benzoquinone, p-chloranilic acid, 2,5-cyclohexadiene-1,4-dione, 2,5-dichloro-3,6-dihydroxy, unii-yj8l3bb7y4, 2,5-dichloro-3,6-dihydroxybenzoquinone, 2,5-dihydroxy-3,6-dichlorobenzoquinone, yj8l3bb7y4, p-quinone, 2,5-dichloro-3,6-dihydroxy, p-benzoquinone, 2,5-dichloro-3,6-dihydroxy |
| InChI Key | IPPWILKGXFOXHO-UHFFFAOYSA-N |
| SMILES | C1(=C(C(=O)C(=C(C1=O)Cl)O)Cl)O |
| IUPAC Name | 2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione |
| Molecular Weight (g/mol) | 208.978 |
| PubChem CID | 66604 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction