missing translation for 'onlineSavingsMsg'
Learn More

Chitin, Practical grade, MP Biomedicals™

Product Code. p-4403646 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
1 kg
Unit Size:
10g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443270 10 g 10g
11463270 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443270 Supplier MP Biomedicals Supplier No. 0210133410

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1398-61-4
Molecular Formula (C8H13NO5)nC16H28N2O11
Synonym chitin from shrimp shells, n-2r-2,4,5-trihydroxy-6-hydroxymethyl oxan-3-yl acetamide, chitin from shrimp shells, practical grade, powder, chitin from shrimp shells, vetec tm reagent grade, chitin from shrimp shells, practical grade, coarse flakes, chitin from shrimp shells, bioreagent, suitable for analysis of chitinase, purified powder

Specifications

Color White to Brown
Quantity 10 g
Formula Weight 770.8
Grade Practical
Physical Form Powder
Chemical Name or Material Chitin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.